Order Entry
Portugal
ContactUsLinkComponent
 
Human recombinant PTP1B (from E. coli)
  ENZOBMLSE3320050
 :  
Human recombinant PTP1B (from E. coli)
  ENZOBMLSE3320050
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    PTP1B
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,5 mg/ml
  • Formulation:
    Liquid. In 50 mM HEPES, pH 7,2, containing 1 mM EDTA, 5 mM DTT, and 0,05% NP-40.

 

 

Produced in E. coli. Human PTP1B (protein tyrosine phosphatase 1B) (aa 1-322).