Order Entry
Portugal
ContactUsLinkComponent
Human recombinant Shp-2 (from E. coli)
Human recombinant Shp-2 (from E. coli)
Catalog # ENZOBMLSE2730020
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Shp-2 (from E. coli)
Catalog # ENZOBMLSE2730020
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Shp-2
  • Formulation:
    Liquid. In 10 mM MES, pH 5,7, containing 100 mM NaCl, 5 mM DTT and 33% glycerol.

Specifications

About this item

Produced in E. coli. full length human Shp-2 (aa 1-593).