Human recombinant nerve growth factor receptor:Fc
Catalog # ENZOALX522044C050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Inhibits rat NGF-mediated activation of rat PC12 cells.
- Reconstitution instructions:Reconstitute in 50 µl sterile water. Further dilutions can be made with medium containing 5% fetal calf serum.
- Protein/peptide name:Nerve Growth Factor receptor:Fc
- Purity:>90% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The cysteine-rich domain of human NGFR (nerve growth factor receptor; p75NTR) (aa 1-250) is fused to the Fc portion of human IgG1.