Order Entry
Portugal
ContactUsLinkComponent
 
Human recombinant Annexin V (from E. coli), APC (Allophycocyanin)
 
Human recombinant Annexin V (from E. coli), APC (Allophycocyanin)

 

  • Conjugation:
    APC (Allophycocyanin)
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Annexin V
  • Purity:
    >89% (SDS-PAGE, HPLC)
  • Formulation:
    Liquid. In 50 mM TRIS, pH 7,4, containing 100 mM sodium chloride, 1% BSA and 0,02% sodium azide.

 

Frequently Bought Together