Rabbit Native Metallothionein-1 (from Liver)
Catalog # ENZOALX202072M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Native
- Source:Liver
- Species:Rabbit
- Seq:02
- Protein/peptide name:Metallothionein-1
- Purity:>95%
- Formulation:Lyophilised
- Purification:Metal content (Cd and Zn): ~9%
Specifications
About this item
Isolated from rabbit liver. Mixture of MT isoforms. Contains mainly MT-1a and MT-2e and a minor portion of MT-2d.
Metallothionein-1 containing zinc and cadmium ions. Suitable for biochemical studies. For cell culture studies, please use Prod. No. ALX-202-070.
Ordering information: Quantity: 1mg contains 0.75 mg of the protein.