Order Entry
Portugal
ContactUsLinkComponent
Human recombinant HSP40, HIS tag
Human recombinant HSP40, HIS tag
Catalog # ENZOALX201274C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant HSP40, HIS tag
Catalog # ENZOALX201274C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    HSP40
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 20 mM TRIS, pH 8,0, containing 2 mM DDT, 0,2 mM PMSF and 10% glycerol.

Specifications

About this item

Produced in E.coli and fused to a His-tag.