Order Entry
Portugal
ContactUsLinkComponent
Mouse recombinant PARP2 (from insect cells (Sf9)), HIS Tag
Mouse recombinant PARP2 (from insect cells (Sf9)), HIS Tag
Catalog # ENZOALX201251C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant PARP2 (from insect cells (Sf9)), HIS Tag
Catalog # ENZOALX201251C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Mouse
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    PARP2
  • Purity:
    >98% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 100 mM sodium chloride, 50 mM imidazole, 0,2% NP-40 and 10% glycerol.
  • Grade:
    High purity

Specifications

About this item

Produced in Sf21 cells. Mouse full-length PARP-2 is fused to a His-tag.