Recombinant DsbA (from E. coli)
Catalog # ENZOALX201153C025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Seq:02
- Protein/peptide name:DsbA
- Purity:>99%
- Concentration:1 mg/ml
- Formulation:Liquid. In water.
Specifications
About this item
Produced in E. coli.