Order Entry
Portugal
ContactUsLinkComponent
Human recombinant SuperKillerTRAIL™ (soluble) (from E. coli)
Human recombinant SuperKillerTRAIL™ (soluble) (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Human recombinant SuperKillerTRAIL™ (soluble) (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    SuperKillerTRAIL™ (soluble)
  • Biological activity:
    Induces apoptosis at concentrations of >5 ng/ml. Note: Does not require a cross-linking enhancer for its potent biological activity. Shows superior activity on human cell lines that require extensive cross-linking of TRAIL-Rs for killing (e.g. Jurkat).
  • Purity:
    >98% (SDS-PAGE)
  • Concentration:
    0,5 mg/ml
  • Formulation:
    Liquid. In 20 mM HEPES, pH 7,9, containing 120 mM NaCl, 0,01% Tween 20, 1% sucrose, 1 mM DTT.
  • Purification:
    Affinity purified. Sterile filtered

Specifications

Frequently Bought Together