Human recombinant Leptin (from E. coli)
Catalog # ENZOALX201034M001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Biological activity:Induces proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
- Reconstitution instructions:Reconstitute with sterile water or 0,4% sodium bicarbonate, pH 8 - 9. Do not reconstitute to less than 0,1 mg/ml. Further dilutions should be made with medium containing 0,1% HSA or BSA.
- Protein/peptide name:Leptin
- Purity:>98% (SDS-PAGE)
- Formulation:Lyophilised from 0,0045 mM sodium bicarbonate.
Specifications
About this item
Produced in E. coli. Homologous to human serum leptin.
Ordering information: Manufactured by BioVendor.