Synthetic melittin
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
PLA2 stimulatory bee venom peptide. Sequence is similar to Phospholipase A2 Activating Peptide (Prod. No. ALX-165-007). Activates PLA2 (5-fold at 10 µg/ml) in BC3H1 cells. For additional technical information see melittin (natural) (Prod. No. ALX-162-006).
- PLA2 activator
Specifications
- Protein/peptide type:Synthetic
- Protein/peptide name:Melittin
- Purity:-