Human EGF receptor (652-657)
Catalog # ENZOALX155023M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Species:Human
- Seq:02
- Protein/peptide name:EGF receptor (652-657)
- Purity:>97%
Specifications
About this item
Specific PKC substrate
Specific substrate (aa 652-657 of mature EGF receptor, without signal peptide (aa 1-24)) for protein kinase C (PKC).