Human;Pig;Rat Endothelin 3
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Ordering information: Quantity: *Sold as net peptide weight.
Specifications
- Species:PigHumanRat
- Protein/peptide name:Endothelin 3
- Purity:>98% (HPLC)
- Formulation:Lyophilised
Specifications
Frequently Bought Together