Dog;Human;Pig Endothelin 2
Catalog # ENZOALX155002PC01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:0,1 mg
- Species:PigHumanDog
- Seq:02
- Protein/peptide name:Endothelin 2
- Purity:>98%
Specifications
About this item
Ordering information: Quantity: *Sold as net peptide weight.