Order Entry
Portugal
ContactUsLinkComponent
 
LH ELISA kit, grow cells
  ENZOENZKIT1070001
 :  
LH ELISA kit, grow cells
  ENZOENZKIT1070001
 :  

 

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Description:
    LH ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    5,2 mIU/ml (1,2 - 280 mIU/ml)

 

 

Highly sensitive LH ELISA kit enabling detection of LH in serum, plasma and tissue culture media in just 2 hours.

  • Sensitive measurement of LH, detecting as little as 5,2 mIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in two hours for up to 39 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of LH in tissue culture media, serum and plasma in human and rat samples.

The LH ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in two hours.

 : Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution.