HSP27 (human), ELISA Kit
Catalog # ENZOADIEKS500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Description:HSP27 (human), ELISA Kit
- Seq:02
- Sensitivity:0,39 ng/ml (range 0,39 - 25 ng/ml)
Specifications
About this item
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of human HSP27 in cell lysates, plasma, serum, and tissue.
The HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Highly sensitive quantitative determination of Hsp27 independent of phosphorylation state.
Ordering information: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.