Order Entry
Portugal
ContactUsLinkComponent
 
Anti-ESR Rabbit Polyclonal Antibody
  ENZOADIVAPSV003E
Anti-ESR Rabbit Polyclonal Antibody
  ENZOADIVAPSV003E

 

  • Antibody type:
    Primary
  • Antigen name:
    Estrogen Receptor
  • Antigen symbol:
    ESR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Xenopus,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Epitope:
    near the C-Terminus
  • Form:
    Liquid
  • Antigen synonyms:
    estrogen receptor alpha delta 4*,5,6,7*/654 isoform|estradiol receptor|nuclear receptor subfamily 3 group A member 1|estrogen receptor|ER-alpha|estrogen receptor alpha delta 3*,4,5,6,7*/819-2 isoform|estrogen receptor alpha delta 4 +49 isoform
  • Storage buffer:
    Borate buffered saline, pH 8,2 containing 25 mM sodium borate, 100 mM boric acid, 75 mM sodium chloride, and 5 mM EDTA
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthetic peptide corresponding to the sequence near the C-terminus of rat CSP. The sequence is completely conserved in bovine.
  • Purification:
    purified by Immunoaffinity chromatography
  • Pk:
    100 µG

 

 

Cysteine string protein (CSP) is a 34 kDa rat brain protein localized to the cytoplasmic surface of the synaptic vesicle. CSP is believed to perform similar functions in the assembly of synaptic vesicle proteins which are essential to membrane trafficking.

Type: Primary
Antigen: ESR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Xenopus, Bovine