Anti-GRPE Rabbit Polyclonal Antibody
ENZOADISPA240J
- Antibody type:Primary
- Antigen symbol:GRPE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Bacteria
- Western blot:Yes
- Seq:02
- Form:Liquid
- Storage buffer:PBS containing 50% glycerol and 0,09% sodium azide
- Storage temperature:−20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Purified E. coli GrpE
- Purification:Purified by Protein A affinity chromatography
- Pk:1 mg
GrpE (Glucose Regulated Protein E. coli) is encoded by a monocistronic E. coli gene, which is under the control of both s32 and s70 transcription factors. The GrpE protein is a so-called co-chaperone because it is known to assist the DnaK (Hsp70) protein to effectively carry out DnaK-dependent chaperone activity (i.e. protein folding, protein transport, disaggregation of heat inactivated proteins, activation of mutant protein).
Type: Primary
Antigen: GrpE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria