Anti-IRAK-4 Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPST206E
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:IRAK-4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Epitope:Near the C-Terminus
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide
- Storage temperature:4 °C
- Shipping temperature:Blue ice not frozen
- Immunogen:Synthetic peptide corresponding to the sequence near the C-terminus of human IRAK-4
- Purification:Purified by peptide affinity chromatography
- Pk:100 µG
Specifications
About this item
IRAK (IL-1 associated serine/threonine kinase) is a critical regulator of IL-1 induced activation of the NF-κB pathway. IL-1 stimulates the association of IRAK with the heterodimer of IL-IRI and IL-IRAcP via the adapter protein MyD88. In addition to its role in IL-1 signaling, IRAK is a component of the signal transduction pathway through which TNF-R1 activates NF-κB dependent gene expression.
Type: Primary
Antigen: IRAK-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Human