Order Entry
Portugal
ContactUsLinkComponent
Anti-LAG3 Mouse Monoclonal Antibody
Anti-LAG3 Mouse Monoclonal Antibody
Catalog # ENZOALX804806C100
Supplier:  ENZO LIFE SCIENCES
undefined
Anti-LAG3 Mouse Monoclonal Antibody
Catalog # ENZOALX804806C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lymphocyte-activation gene 3
  • Antigen symbol:
    LAG3
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 10% glycerol and 0,02% sodium azide
  • Concentration:
    1 mg/ml
  • Purification:
    ≥95% (SDS-PAGE)
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The lymphocyte activation gene-3 (LAG-3, CD223), a member of the immunoglobulin superfamily (IgSF) related to CD4, binds to the major histocompatibility complex (MHC) class II molecules but with higher affinity than CD4. Several alternative mRNA splice-variants of human LAG-3 have been described, two of them encoding potential secreted forms: LAG-3V1 (i.e. the D1-D2 domains of the protein, 36 kDa) and LAG-3V3 (D1-D3, 52 kDa). The longer form was detected by ELISA in the serum of healthy individuals as well as of tuberculosis patients with a favorable outcome. LAG-3 expression by T cell clones correlated with IFN-γ production, and hence soluble LAG-3 has been suggested as a serological marker of Th1 responses.

  • For Immunocytochemistry, Western Blot

Recognizes the 30 aa extra-loop of the first N-terminal D1 domain of human LAG-3.

Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human