Order Entry
Portugal
ContactUsLinkComponent
 
Anti-XRCC1 Rabbit Polyclonal Antibody
  ENZOALX210539R100
Anti-XRCC1 Rabbit Polyclonal Antibody
  ENZOALX210539R100

 

  • Antibody type:
    Primary
  • Antigen name:
    X-ray Repair Complementing Defective Repair in Chinese cells 1
  • Antigen symbol:
    XRCC1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. Antiserum containing 0,1% sodium azide.
  • Antigen synonyms:
    complementing defective|X-ray-repair|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1|repair in Chinese hamster
  • Pk:
    100 µl

 

 

Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human XRCC1.

Type: Primary
Antigen: XRCC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human