Order Entry
Portugal
ContactUsLinkComponent
 
Anti-SYN2 Rabbit Polyclonal Antibody
  ENZOADIVASSV061E
Anti-SYN2 Rabbit Polyclonal Antibody
  ENZOADIVASSV061E

 

  • Antibody type:
    Primary
  • Antigen name:
    Synapsin 2
  • Antigen symbol:
    SYN2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
  • Antigen synonyms:
    Synapsin 2|Synapsin II|Syn2
  • Purification:
    Protein A affinity purified
  • Pk:
    100 µG

 

 

Synapsin I, II and III belong to a family of neuron-specific phosphoproteins which maintain a reserve pool of vesicles in the vicinity of the active zone by tethering synaptic vesicles to each other and to an Actin-based cytoskeleton meshwork. They have been shown to regulate neurotransmitter release in mature synapses and to accelerate development of the nervous system.

  • For Immunoprecipitation, Western Blot

Recognizes mouse and rat synapsin II. Detects a band of ~74kDa (synapsin Iia) and ~54kDa (synapsin Iib) by Western blot.

Type: Primary
Antigen: SYN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse