Anti-HSF1 Rabbit Polyclonal Antibody
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.
- For Western Blot
Recognizes human HSF1. Predicted MW of ~60kDa. Detects a band of 85-95kDa by Western blot depending on hyperphosphorylation of the native protein.
Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
- Catalog No:
- ENZOADISPA902F
- ENZOADISPA902D
- Antigen symbol:
- HSF1
- HSF1
- Antigen name:
- Heat Shock Transcription Factor 1
- Heat Shock Transcription Factor 1
- Antigen synonyms:
- HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Form:
- Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
- Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
- Purification:
- Protein A affinity purified
- Protein A affinity purified
Frequently Bought Together