Order Entry
Portugal
ContactUsLinkComponent
Anti-liver FABP Polyclonal Antibody
Anti-liver FABP Polyclonal Antibody
Catalog # BSBTA02477
Supplier:  Boster Bio
undefined
Anti-liver FABP Polyclonal Antibody
Catalog # BSBTA02477
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    liver FABP
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for liver FABP detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: liver FABP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat