NADPH regeneration system
Catalog # PROMV9510
Supplier: Promega
CAS Number:
Specifications
- Pk:1.000 Assays
- Description:NADPH Regeneration System
Specifications
About this item
NADPH regeneration system is an enzyme substrate combination that reduces NADP+ to NADPH.
- A component of the P450-Glo CYP450 screening systems
- This system consists of two reagents, Solution A and Solution B
Ordering information: Solution A contains the substrate NADP+ and glucose-6-phosphate and is supplied as a 20X concentrate. Solution B contains the enzyme glucose-6-phosphate dehydrogenase at a 100X concentration. These system should be ordered separately.