Human recombinant Arginase 1 (from E. coli)
Catalog # PRSI92-205
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Arginase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:35.8 kD
- Sequence:Met1-lys322
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM TrisHCl, pH 7.5 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
ARG1 is a member of the ureohydrolase family of enzymes. ARG1 can catalyse the hydrolysis of arginine to ornithine and urea. In the urea cycle, ARG1 catalyses the fifth and final step, a series of biochemical reactions in mammals during which the body disposes of harmful ammonia. ARG1 is a cytosolic enzyme and expressed widely in the liver as part of the urea cycle. Inherited deficiency of this ARG1 causes argininemia, which is an autosomal recessive disorder characterised by hyperammonemia.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.