Human recombinant tripeptidyl-peptidase I (from cells)
Catalog # PRSI91-745
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Tripeptidyl-Peptidase I
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:60.35 kD
- Sequence:Ser20-Pro563
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM GaCl₂, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Tripeptidyl-Peptidase 1 (TPP1) belongs to the peptidase S53 family. TPP1 is detected in all tissues examined with highest levels in heart and placenta and relatively similar levels in other tissues. TPP1 is lysosomal serine protease with tripeptidyl-peptidase I activity. TPP1 may act as a non-specific lysosomal peptidase which generates tripeptides from the breakdown products produced by lysosomal proteinases. TPP1 requires substrates with an unsubstituted N-terminus. TPP1 mutations have also been shown to cause neuronal ceroid lipofuscinosis type 2 (CLN2).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.