Human recombinant NHP2 (from E. coli)
Catalog # PRSI92-211
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:H/ACA ribonucleoprotein complex subunit 2|NHP2P.|NHP2|NOLA2|snoRNP protein NHP2|Nucleolar protein family A member 2
- Protein/peptide name:NHP2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:19.3 kD
- Sequence:Met1-Leu153
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris,100mM NaCl,1mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
NHP2 belongs to the H/ACA snoRNPs gene family. snoRNPs are involved in various aspects of rRNA processing and modification and have been organised into 2 families: C/D and H/ACA. NHP2 forms a small ribonucleoprotein particle with GAR1 (NOLA1). NHP2 is involved in a variety of aspects of rRNA processing and modification. NHP2 localises to the dense fibrillar component of the nucleolus and in nuclear Cajal bodies. NHP2 may also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.