Human recombinant heat shock beta 7 (from E. coli)
Catalog # PRSI92-207
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Pregnancy-specific beta-1 glycoprotein E|Pregnancy specific beta-1-glycoprotein 2|pregnancy-specific beta-1-glycoprotein 2|pregnancy-specific beta-1-glycoprotein 7|PSBG2|HSPB7|PSG2
- Protein/peptide name:Heat Shock beta 7
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:19.6 kD
- Sequence:Met1-Ile170
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris,0.2M NaCl,2mM DTT,50%glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
PSG2 is a secreted protein that in humans is encoded by the PSG2 gene. It is a member of the human pregnancy-specific glycoproteins (PSGs) family. These proteins are synthesised in large amounts by placental trophoblasts and released into the maternal circulation during pregnancy. PSG2 consist of a single N domain, with structural similarity to the immunoglobulin variable domains, followed by a variable number of immunoglobulin constant-like A and/or B domains. It has an arg-gly-asp (RGD) motif, which has been shown to function as an adhesion recognition signal for several integrins, in the N-terminal domain.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.