Human recombinant INPP1 (from cells)
Catalog # PRSI91-702
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Inositol Polyphosphate 1-Phosphatase|INPP1|IPPase|IPP
- Protein/peptide name:INPP1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:45 kD
- Sequence:Met1-Thr399
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Inositol Polyphosphate 1-Phosphatase (INPP1) is a member of the inositol monophosphatase family. INPP1 is widely expressed in tissues, with highest expression levels observed in the pancreas and kidney. INPP1 is inhibited by Li+. Removing the phosphate group at position 1 of the inositol ring from the polyphosphates inositol 1,4-bisphosphate and inositol 1,3,4-trisphophosphate. INPP1 is involved in signal transduction and phosphatidylinositol signaling pathway.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag