Order Entry
Portugal
ContactUsLinkComponent
Human recombinant transthyretin (from cells)
Human recombinant transthyretin (from cells)
Catalog # PRSI91-495
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant transthyretin (from cells)
Catalog # PRSI91-495
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Prealbumin|TBPA|TTR|Transthyretin|PALB|ATTR
  • Protein/peptide name:
    Transthyretin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.8 kD
  • Sequence:
    Gly21-Glu147
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Transthyretin is a secreted and cytoplasm protein which belongs to the Transthyretin family. Transthyretin is detected in serum and cerebrospinal fluid (at protein level). It is highly expressed in choroid plexus epithelial cells. It is also detected in retina pigment epithelium and liver. Each monomer of Transthyretin has two 4-stranded beta sheets and the shape of a prolate ellipsoid. Antiparallel beta-sheet interactions link monomers into dimers. A short loop from each monomer forms the main dimer-dimer interaction. These two pairs of loops separate the opposed, convex beta-sheets of the dimers to form an internal channel. Defects in Transthyretin are the cause of amyloidosis type 1 (AMYL1) which is a hereditary generalized amyloidosis due to transthyretin amyloid deposition. Protein fibrils can form in different tissues leading to amyloid polyneuropathies, amyloidotic cardiomyopathy, carpal tunnel syndrome, systemic senile amyloidosis. The disease includes leptomeningeal amyloidosis that is characterized by primary involvement of the central nervous system.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag