Order Entry
Portugal
ContactUsLinkComponent
Human recombinant FGF R5 (from cells)
Human recombinant FGF R5 (from cells)
Catalog # PRSI91-471
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FGF R5 (from cells)
Catalog # PRSI91-471
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Fibroblast Growth Factor Receptor 5|FGFR5|FGFR-like Protein|FGF Homologous Factor Receptor|FGF Receptor-like Protein 1|FGFR-5|FHFR|FGFRL1|Fibroblast Growth Factor Receptor-like 1
  • Protein/peptide name:
    FGF R5
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    39.9 kD
  • Sequence:
    Ala25-Pro378
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Fibroblast Growth Factor Receptor-Like 1 (FGFRL1) is a single-pass type I membrane protein that belongs to the FGF receptor family. The mature human FGFRL1 consists of a 354 amino acid extracellular domain (ECD) with 3 Ig-like C2-type domains, a 21 amino acid transmembrane segment, and a 134 amino acid cytoplasmic domain. FGFR1 expressed in various tissues, preferentially in cartilaginous tissues and pancreas. It highly expressed in the liver, kidney, heart, brain and skeletal muscle, weakly expressed in the lung, small intestine and spleen. FGFRL1 has a negative effect on cell proliferation.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag