- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:PRKCE|PKCE|nPKC-Epsilon|Protein Kinase C Epsilon Type
- Protein/peptide name:PKC epsilon
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:19.64 kD
- Sequence:Gln580-Pro737
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological assays
Protein Kinase C Epsilon type is a member of the serine- and threonine-specific protein kinase family that can be activated by calcium and the second messenger diacylglycerol. Protein Kinase C Epsilon contains these domains: one AGC-kinase C-terminal domain, one C2 domain, one protein kinase domain and two phorbol-ester/DAG-type zinc fingers. Protein Kinase C Epsilon phosphorylate a variety of protein targets and has been identified to participate in diverse cellular signaling pathways. It has many different cellular functions, such as neuron channel activation, apoptosis, cardioprotection from ischemia, heat shock response, as well as insulin exocytosis. Protein Kinase C Epsilon also serves as the receptor for phorbol esters, a class of tumor promoters.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag