Human recombinant SCARB2 (from HEK293 cells)
Catalog # PRSI96-674
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized rh SCARB2 / CD36L2/ LIMP2 Fc Chimera at 5 μg/mL (100 μl/well) can bind rhTSP-2/His with a linear range of 0. 1 - 5 μg/mL.
- Protein synonyms:EPM4|CD36L2|LIMP2|CD36|AMRF|LIMPII|SR-BII|LGP85|SCARB2|HLGP85
- Protein/peptide name:SCARB2
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:72.5 kDa
- Formulation:Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
- Tested applications:Western Blot
Specifications
About this item
Scavenger receptor class B member 2 (SCARB2) is also known as Lysosome membrane protein 2 (LIMP2), 85 kDa lysosomal membrane sialoglycoprotein (LGP85), CD36 antigen-like 2 (CD36L2, LIMP2), which belongs to the CD36 family. SCARB2 acts as a lysosomal receptor for glucosylceramidase (GBA) targeting.