Human recombinant S100P (from E. coli)
Catalog # PRSI96-671
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:S100P|MIG9|S100E
- Protein/peptide name:S100P
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:10.4 kDa
- Formulation:Lyophilized, 50 mM Tris,150 mM Nacl,pH8.0
- Tested applications:Western Blot
Specifications
About this item
S100 calcium-binding protein P (S100P),a member of the S-100 family, is also known as protein S100-P, migration-inducing gene 9 protein (MIG9) and protein S100-E,which contains two EF-hand domains. As for subunit structure,S100P is homodimer and heterodimer with S100A1. S100P may function as calcium sensor and contribute to cellular calcium signaling. In a calcium-dependent manner, S100P functions by interacting with other proteins, such as EZR and PPP5C, and indirectly plays a role in physiological processes like the formation of microvilli in epithelial cells. Furthermore,S100P may stimulate cell proliferation in an autocrine manner via activation of the receptor for activated glycation end products (RAGE).