Human recombinant IFNGR1 (from HEK293 cells)
Catalog # PRSI96-406
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:200 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:IFNGR|CD119|CDw119|IFN-gamma-R1
- Protein/peptide name:IFNGR1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:52.4 kDa
- Formulation:Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
- Tested applications:Western Blot
Specifications
About this item
Interferon gamma receptor 1 (IFNGR1) is also known as CD antigen CD119, which belongs to the type II cytokine receptor family. IFNGR1 contains two fibronectin type-III domains and two Ig-like C2-type (immunoglobulin-like) domains. IFNGR1 / CD119 is receptor for interferon gamma. Two receptors bind one interferon gamma (IFNG) dimer.