Human recombinant ANGPTL4 (from HEK293 cells)
Catálogo # PRSI96-028
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:PSEC0166|Angiopoietin-related protein 4|ANGPTL4|Hepatic fibrinogen/angiopoietin-related protein|ARP4|HFARP|PP1158|FIAF|PGAR|UNQ171/PRO197
- Protein/peptide name:ANGPTL4
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:27.9 kDa
- Sequence:166-406
- Formulation:Lyophilized, 50 mM MOPS, 100 mM NaCl, pH7.2 with 0.1% CHAPS
- Tested applications:Western Blot
Specifications
About this item
Angiopoietin-related protein 4 (ANGPTL4) is also known as Hepatic fibrinogen / angiopoietin - related protein (HFARP), ARP4, PGAR. ANGPTL4 contains one fibrinogen C-terminal domain. ANGPTL4 is a homooligomer , and the homooligomer undergoes proteolytic processing to release its carboxyl fibrinogen-like domain, which circulates as a monomer. The homooligomer unprocessed form is able to interact with the extracellular matrix. ANGPTL4 may act as a regulator of angiogenesis and modulate tumorigenesis. ANGPTL4 inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 may exert a protective function on endothelial cells through an endocrine action.