Human recombinant MCP1 (from E. coli)
Catalog # PRSI40-564
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Biological activity was determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/mL. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
- Protein synonyms:HC11|MCAF|SMC-CF|GDCF-2|HSMCR30|SCYA2|MCP1|MCP-1|C-C motif chemokine 2
- Protein/peptide name:MCP1
- Purity:>98%
- Formulation:Lyophilized
Specifications
About this item
The MCP proteins belong to the CC chemokine family, and signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross reacts across species. Recombinant human MCP-1 is an 8.6 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
- Store in working aliquots at -20°C. Avoid freeze/thaw cycles.