Order Entry
Portugal
ContactUsLinkComponent
 
Human TCTEX1D4 peptide (19 aa near N-terminus)
  PRSI6957P
 :  
undefined
Human TCTEX1D4 peptide (19 aa near N-terminus)
  PRSI6957P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa near N-terminus
  • Protein/peptide name:
    TCTEX1D4 Peptide (19 aa near N-terminus)
  • Tested applications:
    Blocking

 

 

TCTEX1D4 peptide is used for blocking the activity of TCTEX1D4 antibody.