Order Entry
Portugal
ContactUsLinkComponent
 
Anti-TP1 Rabbit Polyclonal Antibody
Catalog # PRSI2345
Supplier:  ProSci Inc.
undefined
Anti-TP1 Rabbit Polyclonal Antibody
Catalog # PRSI2345
Supplier:  ProSci Inc.

Specifications

  • Antigen symbol:
    TP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    TP1 antibody was raised against a 20 amino acid synthetic peptide from near the amino terminus of human TP1.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

TP1 Antibody: Telomerase is an RNA-dependent DNA polymerase that uses an RNA component to add telomeric repeat sequences at the ends of chromosomes. Besides the RNA component which serves as the template that specifies the telomeric repeat, the telomerase complex contains a reverse transcriptase protein (TRT) and various accessory proteins including the telomerase-associated protein 1 (TP1). Telomerase activity is low in most somatic cells, causing the gradual shortening of telomeres which can ultimately lead to telomere fusion and cell death. High levels of telomerase activity are widely seen in cancerous cells and while recent experiments have suggested that telomerase may be a viable target in cancer therapy, expression levels of TP1 do not correlate with malignancy. At least two isoforms of TP1 are known to exist.

Type:
Antigen: TP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat