Especificações
- Antigen name:Tripartite Motif Containing 5 alpha
- Antigen symbol:TRIM5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:TRIM5 alpha antibody was raised against a synthetic peptide corresponding to amino acids near the C-terminus of rhesus monkey TRIM5 alpha.
- Tested applications:E, WB, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Especificações
Sobre este item
TRIM5 alpha Antibody: TRIM5 is a member of a broad family of otherwise unrelated proteins defined by the presence of a tripartite motif containing a RING domain, a B-box type 1, and a B-box type 2, followed by a coiled-coil region. TRIM5 has six alternately spliced isoforms, the longest of which is the alpha variant which also contains a carboxy-terminal B30.2 (SPRY) domain. Expression of TRIM5 alpha variants from humans, rhesus monkeys, and African green monkeys enabled resistance to infection by various retroviruses including HIV-1, albeit at differing efficiencies. All TRIM5 alpha variants could inhibit at least two different retroviruses, but not from those viruses isolated from the same species, suggesting that TRIM5 alpha acts as a natural barrier to cross-species retrovirus transmission.
Type:
Antigen: TRIM5 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human