Anti-TRFP / MED20 Rabbit Polyclonal Antibody
BSBTA09407-1
- Antibody type:Primary
- Antigen symbol:TRFP / MED20
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for TRFP / MED20 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: TRFP / MED20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat