Fluorometric TUNEL system, DeadEnd™
Catalog # PROMG32500060
Supplier: Promega
CAS Number:
Specifications
- Pk:60 Tests
- Description:DeadEnd™ Fluorometric TUNEL system
Specifications
About this item
The DeadEnd™ Fluorometric TUNEL system is a classic TUNEL (TdT-mediated dUTP Nick-End Labeling) assay designed for the specific detection and quantitation of apoptotic cells within a cultured cells or FFPE tissue sections.
- Simple and accurate
- Plastic coverslips provided simplify sample handling
- Direct incorporation of fluorescent nucleotide reduces number of incubation steps
- System provides sufficient reagents for 60 assays of 50 μl each
The DeadEnd™ Fluorometric TUNEL System measures the fragmented DNA of apoptotic cells by catalytically incorporating fluorescein-12-dUTP at 3´-OH DNA ends using Terminal Deoxynucleotidyl Transferase (TdT), which forms a polymeric tail using the principle of the TUNEL assay. The fluorescein-12-dUTP-labeled DNA can then be visualised directly by fluorescence microscopy or quantitated by flow cytometry.
Delivery: Kit includes plastic coverslips, equilibration buffer, nucleotide mix, SSC 20X, terminal deoxynucleotidyl transferase (recombinant) and proteinase K.