Nerve growth factor, 2,5S, murine
PROMG51410100
: Promega
:
- Pk:100 µG
- Description:Murine 2,5S nerve growth factor (2,5S mNGF)
Murine 2,5S nerve growth factor (2,5S mNGF) is a 26 kDa protein composed of two identical 118 amino acid chains.
- Purified from male mouse submaxillary glands (Bocchini and Angeletii method)
- Mediates phosphorylation of specific intracellular proteins
Target cells of this molecule include sympathetic and sensory neurons and derivatives of nerve cells such as adrenal medulla pheochromocytoma (PC12) cells.
2,5S mNGF exhibits an ED50 value below 2 ng/ml using a PC-12 serum-free survival assay.
: Supplied as a sterile-filtered, lyophilised powder.