Order Entry
Portugal
ContactUsLinkComponent
Protective hood, Gridel
Protective hood, Gridel
Catalog # DALLCC85278339-AP
Supplier:  Honeywell Safety
undefined
Protective hood, Gridel
Catalog # DALLCC85278339-AP
Supplier:  Honeywell Safety
Specifications
  • Colour:
    Pink
  • Description:
    Protective hood, made of flame retardant M2
  • Pk:
    20
Specifications
About this item
Gridel protective hood is designed to isolate the wearer's respiratory tracs when working in surroundings with suspected or low air borne contamination.
  • Single use respiratory protection apparatus fed by compressed air
  • Comfortable and lightweight
  • Can be fitted with a tearable safety strip
  • Adjustable air flow
  • 360 °C visual field
  • Various nipples available
Ideal for use in chemical, nuclear, pharmaceutical, oil and gas industries.
Certifications: EN 143, EN 146, EN 270, EN 1073 and EN 1835.