Order Entry
Portugal
Orders LinkContactUsLinkComponent
dsDNA System, QuantiFluor®
dsDNA System, QuantiFluor®
Supplier:   Promega
dsDNA System, QuantiFluor®
Supplier:  Promega
Certificates
dsDNA System, QuantiFluor®
dsDNA System, QuantiFluor®
2 Options Available Below
Product Details & Documents
The QuantiFluor® dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The system is suitable for use in many molecular biology applications, including next-gen sequencing and other techniques where accurate DNA quantitation is important.
  • Specific quantitation of dsDNA
  • More sensitive than other methods for low-concentration samples
QuantiFluor® dsDNA dye shows minimal binding to single stranded DNA (ssDNA), RNA, protein and interfering compounds, allowing specific quantitation of dsDNA.
Delivery information: Kit includes QuantiFluor® dsDNA dye (1×1 ml), Lambda DNA standard (1×100 µg) and 20X TE buffer, pH 7,5 (1×25 ml).
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • PROME2671
    QuantiFluor® dsDNA system, trial size
    QuantiFluor® dsDNA system, trial size
    0,1 mL
    Log in for availability
    Log in for availability
    1 * 0,1 mL
    1 * 0,1 mL - 226,00 €
    226,00 €
    1 * 0,1 mL
       226,00 €
  • PROME2670
    QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale
    QuantiFluor® dsDNA System, sufficient reagents for 2000 assays at a 200 µl scale
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 639,00 €
    639,00 €
    1 * 1 mL
       639,00 €