Order Entry
Portugal
Orders LinkContactUsLinkComponent
RNAgents® Denaturing Solution
RNAgents® Denaturing Solution
RNAgents® Denaturing Solution, 120 mL
Catalog # PROMZ5651
Supplier:   Promega
RNAgents® Denaturing Solution
Catalog # PROMZ5651
Supplier:  Promega
CAS Number:  
RNAgents® Denaturing Solution
RNAgents® Denaturing Solution
RNAgents® Denaturing Solution, 120 mL
CAS Number:  
Price
313,00 €
Log in for availability
1 * 120 mL
Product Details & Documents
RNAgents Denaturing Solution lyses cells or tissue under conditions that rapidly inhibit ribonucleases.
  • Lyses Cells or Tissue Under Conditions that Rapidly Inhibit Ribonucleases
  • Uses two potent inhibitors of RNase, guanidine thiocyanate and β-mercaptoethanol
  • Designed to be used with acidic phenol:chloroform and alcohol (isopropanol) for purification of total RNA
  • Fits RNAgents® Total RNA Isolation System (PAZ5110)
RNAgents Denaturing Solution lyses cells or tissue under conditions that rapidly inhibit ribonucleases, using two potent inhibitors of RNase, guanidine thiocyanate and beta-mercaptoethanol. The RNAgents Denaturing Solution is designed to be used in concert with acidic phenol:chloroform and alcohol (isopropanol) for purification of total RNA.
Specifications
Specifications
  • Pk:
    120 mL
  • Description:
    RNAgents® Denaturing Solution