Order Entry
Portugal
Orders LinkContactUsLinkComponent
Lysis buffer, Glo, 1X

Lysis buffer, Glo, 1X

Glo Lysis Buffer, 1X, 100 ml, 100 mL
Catalog # PROME2661
Supplier:   Promega
Lysis buffer, Glo, 1X
Catalog # PROME2661
Supplier:  Promega
CAS Number:  
Lysis buffer, Glo, 1X
Lysis buffer, Glo, 1X

Glo Lysis Buffer, 1X, 100 ml, 100 mL

CAS Number:  
Price
199,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Glo Lysis Buffer (GLB), 1X, is a proprietary formulation developed to promote rapid lysis (within 5 minutes) of cultured mammalian cells without scraping or performing freeze-thaw cycles.

  • No need for cell scraping or freeze-thaw cycles
  • Compatible with Bright-Glo™, Steady-Glo®, ONE-Glo™ and Renilla-Glo® luciferase assay reagents
  • Firefly luciferase enzyme in Glo Lysis Buffer is stable at room temperature for at least 48 hours
Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Glo Lysis Buffer, 1X, 100 ml

Specifications