Order Entry
Portugal
ContactUsLinkComponent
Cell signaling assays, PDE-Glo™ Phosphodiesterase Assay
Cell signaling assays, PDE-Glo™ Phosphodiesterase Assay
Catalog # PROMV1361
Supplier:  Promega
CAS Number:  
undefined
Cell signaling assays, PDE-Glo™ Phosphodiesterase Assay
Catalog # PROMV1361
Supplier:  Promega
CAS Number:  

Specifications

  • Pk:
    1.000 Assays
  • Description:
    PDE-Glo™ Phosphodiesterase Assay
  • Seq:
    01
  • Tests per kit:
    1000 assays in a 384-well plate format

Specifications

About this item

PDE-Glo™ Phosphodiesterase Assay is a luminescent, high-throughput screening (HTS) method for measuring cyclic nucleotide phosphodiesterase (PDE) activity from purified sources. The PDE-Glo™ Assay is used for screening library compounds to identify inhibitors of either cAMP- or cGMP-specific PDEs. It is optimised to work with both cAMP- and cGMP-dependent phosphodiesterases.

  • Sensitive and easy to use
  • Assay complete in >1 hour after PDE reaction
  • Scalable to 1536-well plate formats
  • Simple luminescence detection

Cyclic nucleotide phosphodiesterases are involved in many cellular processes due to their ability to hydrolyse, and thus control, the levels of the second-messenger signaling molecules cAMP and cGMP. The availability of selective inhibitors for PDEs has facilitated their use as tools to study cyclic nucleotide signaling and paved the way to investigate the role of PDEs in cellular and tissue pathologies.

Delivery: Kit includes PDE-Glo™ reaction buffer (5X), PDE-Glo™ detection buffer (5X), PDE-Glo™ termination buffer (5X), Protein Kinase A, cGMP (1 mM), cAMP (1 mM), Kinase-Glo® substrate, Kinase-Glo® buffer.