Magnetic separation device, MagnaBot® II
PROMV8351
: Promega
- Description:MagnaBot® II magnetic separation device
- Pk:1
These magnetic separation device is designed to work with a 96-well PCR plate. A 96-well PCR plate containing MagneSil® paramagnetic particles is placed on the unit to draw the particles to the side and away from the bottom of each well. This allows the quantitative removal of liquids.